Learn More
Aviva Systems Biology Corp ZNF498 Antibody - middle region (ARP34485_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34485_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF498. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF498. Alternate names: ZSCAN25, ZNF498ApplicationIHC, WB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52960