Learn More
Aviva Systems Biology Corp ZNF498 Antibody - N-terminal region (ARP39845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF498. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF498. Alternate names: ZSCAN25, ZNF498ApplicationWB. Species reactivity: Horse, Human, Rabbit
Catalog No. 50-014-57199