Learn More
Aviva Systems Biology Corp ZNF499 Antibody - middle region (ARP30017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF499. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF499. Alternate names: ZNF499ApplicationIHC, WB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54099