Learn More
Aviva Systems Biology Corp ZNF500 Antibody - middle region (ARP31852_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31852_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF500. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF500. Alternate names: ZKSCAN18ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54340