Learn More
Aviva Systems Biology Corp ZNF502 Antibody - N-terminal region (ARP36100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF502. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF502. Alternate names: FLJ12515, FLJ14855ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-56013