Learn More
Aviva Systems Biology Corp ZNF503 Antibody - middle region (ARP30022_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF503. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF503. Alternate names: FLJ45745, MGC2555, NOLZ-1, NOLZ1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54102