Learn More
Aviva Systems Biology Corp ZNF510 Antibody - middle region (ARP39154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF510. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF510. Alternate names: KIAA0972, MGC33740ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56923