Learn More
Aviva Systems Biology Corp ZNF511 Antibody - middle region (ARP36159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF511. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF511. Alternate names: MGC30006ApplicationWB. Species reactivity: Human
Catalog No. 50-284-484