Learn More
Aviva Systems Biology Corp ZNF511 Antibody - N-terminal region (ARP37809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF511. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF511. Alternate names: MGC30006ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56437