Learn More
Aviva Systems Biology Corp ZNF512B Antibody - middle region (ARP33182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF512B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF512B. Alternate names: GM632, KIAA1196, MGC149845, MGC149846ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54908