Learn More
Aviva Systems Biology Corp ZNF513 Antibody - N-terminal region (ARP31555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF513. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF513. Alternate names: FLJ32203, HMFT0656, RP58ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54263