Learn More
Aviva Systems Biology Corp ZNF514 Antibody - middle region (ARP39724_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39724_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF514. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF514. Alternate names: MGC126229, MGC126230ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-823