Learn More
Aviva Systems Biology Corp ZNF514 Antibody - N-terminal region (ARP36083_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36083_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF514. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF514. Alternate names: MGC126229, MGC126230ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-62990