Learn More
Aviva Systems Biology Corp ZNF528 Antibody - middle region (ARP36066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF528. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF528. Alternate names: KIAA1827, MGC126761, MGC138155ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55999