Learn More
Aviva Systems Biology Corp ZNF540 Antibody - middle region (ARP39918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF540. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF540. Alternate names: DKFZp313K2238, DKFZp547B0714, FLJ16004, Nbla10512ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57231