Learn More
Aviva Systems Biology Corp ZNF541 Antibody - middle region (ARP30068_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30068_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF541. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF541. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52820