Learn More
Aviva Systems Biology Corp ZNF547 Antibody - N-terminal region (ARP34900_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34900_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF547. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF547. Alternate names: FLJ31100ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55604