Learn More
Aviva Systems Biology Corp ZNF550 Antibody - N-terminal region (ARP36214_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF550. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF550. Alternate names: MGC41917ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56072