Learn More
Aviva Systems Biology Corp ZNF555 Antibody - N-terminal region (ARP36204_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36204_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF555. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF555. Alternate names: MGC26707ApplicationIHC, WB. Species reactivity: Dog, Human
Catalog No. 50-014-56064