Learn More
Aviva Systems Biology Corp ZNF556 Antibody - N-terminal region (ARP36049_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36049_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF556. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF556. Alternate names: FLJ11637ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-55991