Learn More
Aviva Systems Biology Corp ZNF559 Antibody - N-terminal region (ARP36069_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36069_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF559. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF559. Alternate names: NBLA00121ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56001