Learn More
Aviva Systems Biology Corp ZNF564 Antibody - N-terminal region (ARP36149_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36149_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF564. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF564. Alternate names: MGC26914ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-56032