Learn More
Aviva Systems Biology Corp ZNF566 Antibody - middle region (ARP36086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF566. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF566. Alternate names: FLJ14779, MGC12515ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-56007