Learn More
Aviva Systems Biology Corp ZNF567 Antibody - N-terminal region (ARP39913_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39913_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF567. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF567. Alternate names: MGC45586ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57230