Learn More
Aviva Systems Biology Corp ZNF569 Antibody - N-terminal region (ARP39902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF569. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF569. Alternate names: FLJ32053, ZAP1, ZNFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57226