Learn More
Aviva Systems Biology Corp ZNF57 Antibody - C-terminal region (ARP39986_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39986_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF57. Alternate names: ZNF424ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-57254