Learn More
Aviva Systems Biology Corp ZNF57 Antibody - N-terminal region (ARP39985_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39985_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF57. Alternate names: ZNF424ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53167