Learn More
Aviva Systems Biology Corp ZNF571 Antibody - N-terminal region (ARP35923_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35923_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF571. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF571. Alternate names: HSPC059, MGC138690ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55927