Learn More
Aviva Systems Biology Corp ZNF572 Antibody - N-terminal region (ARP39892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF572. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF572. Alternate names: FLJ38002ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57221