Learn More
Aviva Systems Biology Corp ZNF575 Antibody - middle region (ARP34490_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF575. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF575. Alternate names: FLJ32567ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55416