Learn More
Aviva Systems Biology Corp ZNF575 Antibody - N-terminal region (ARP40009_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF575. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF575. Alternate names: FLJ32567ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig
Catalog No. 50-014-57260