Learn More
Aviva Systems Biology Corp ZNF577 Antibody - C-terminal region (ARP36077_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36077_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF577. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF577. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56004