Learn More
Aviva Systems Biology Corp ZNF579 Antibody - middle region (ARP39910_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39910_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF579. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF579. Alternate names: FLJ35453ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57229