Learn More
Aviva Systems Biology Corp ZNF579 Antibody - C-terminal region (ARP36193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF579. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF579. Alternate names: FLJ35453ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-284-485