Learn More
Aviva Systems Biology Corp ZNF582 Antibody - N-terminal region (ARP36145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF582. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF582. Alternate names: FLJ30927ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-014-56029