Learn More
Aviva Systems Biology Corp ZNF596 Antibody - C-terminal region (ARP36232_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36232_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF596. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF596. Alternate names: FLJ36123ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Pig, Rabbit, Zebrafish
Catalog No. 50-014-56080