Learn More
Aviva Systems Biology Corp ZNF597 Antibody - N-terminal region (ARP39896_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39896_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF597. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF597. Alternate names: ApplicationWB. Species reactivity: Horse, Human, Rabbit
Catalog No. 50-014-57223