Learn More
Aviva Systems Biology Corp ZNF597 Antibody - middle region (ARP39897_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF597. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF597. Alternate names: FLJ33071ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-57224