Learn More
Aviva Systems Biology Corp ZNF598 Antibody - middle region (ARP37823_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37823_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF598. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF598. Alternate names: DKFZp762F135, FLJ00086ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56442