Learn More
Aviva Systems Biology Corp ZNF599 Antibody - N-terminal region (ARP50349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF599. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF599. Alternate names: -ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-285-506