Learn More
Aviva Systems Biology Corp ZNF607 Antibody - middle region (ARP36078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF607. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF607. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56005