Learn More
Aviva Systems Biology Corp ZNF608 Antibody - middle region (ARP32951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF608. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF608. Alternate names: DKFZp781C0723, MGC166851, NY-REN-36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54829