Learn More
Aviva Systems Biology Corp ZNF609 Antibody - N-terminal region (ARP31708_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31708_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF609. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF609. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54315