Learn More
Aviva Systems Biology Corp ZNF613 Antibody - N-terminal region (ARP39628_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39628_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF613. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF613. Alternate names: FLJ13590ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-57118