Learn More
Aviva Systems Biology Corp ZNF614 Antibody - middle region (ARP39641_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39641_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF614. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF614. Alternate names: FLJ21941, MGC120638ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-57121