Learn More
Aviva Systems Biology Corp ZNF620 Antibody - N-terminal region (ARP50835_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50835_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF620. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF620. Alternate names: MGC50836ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-60446