Learn More
Aviva Systems Biology Corp ZNF624 Antibody - N-terminal region (ARP50646_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF624. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF624. Alternate names: KIAA1349, MGC119602, MGC119603, MGC119605ApplicationWB. Species reactivity: Human
Catalog No. 50-285-524