Learn More
Aviva Systems Biology Corp ZNF627 Antibody - N-terminal region (ARP34806_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34806_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF627. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF627. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55570