Learn More
Aviva Systems Biology Corp ZNF628 Antibody - middle region (ARP50733_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF628. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF628. Alternate names: MGC105139, ZEC, Zfp628ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-535