Learn More
Aviva Systems Biology Corp ZNF630 Antibody - middle region (ARP40092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF630. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF630. Alternate names: FLJ20573, MGC138344, dJ54B20.2ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-57292